missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanemethylamine, 98+%

Catalog No. AAL1465903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1465903 1 g
AAL1465906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1465903 Supplier Thermo Scientific Chemicals Supplier No. L1465903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4448-77-5 | C8H17N | 127.231 g/mol

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix with water.

Notes
Air Sensitive, Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 4448-77-5
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00021674
InChI Key CAOQEOHEZKVYOJ-UHFFFAOYSA-N
Synonym cycloheptanemethylamine, cycloheptylmethylamine, cycloheptanemethanamine, acmc-20agqt, cycloheptylmethyl amine, cycloheptane methylamine, aminomethyl cycloheptane, n-cycloheptylmethyl-amine, aminomethylcycloheptane
PubChem CID 78194
IUPAC Name cycloheptylmethanamine
SMILES C1CCCC(CC1)CN

Specifications

Density 0.884
Boiling Point 98°C to 101°C (35 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.474
Quantity 1 g
UN Number UN2735
Beilstein 2958734
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 127.23
Percent Purity ≥98%
Chemical Name or Material Cycloheptanemethylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 224-692-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only