missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptane, 99%

Catalog No. AC111011000
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111011000 100 mL Glass bottle
AC111010050 5 mL Glass bottle
AC111010250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111011000 Supplier Thermo Scientific Chemicals Supplier No. 111011000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 291-64-5 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 291-64-5
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00004147
InChI Key DMEGYFMYUHOHGS-UHFFFAOYSA-N
Synonym heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
PubChem CID 9265
IUPAC Name cycloheptane
SMILES C1CCCCCC1

Specifications

Melting Point -12°C
Color Colorless
Density 0.8100g/mL
Boiling Point 118.4°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.446
Linear Formula C7H14
Quantity 100 mL
Beilstein 05,29
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 98.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cycloheptane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-030-2

RUO – Research Use Only