missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptane, 99%

Catalog No. AAB2290206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2290206 5 g
AAB2290214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2290206 Supplier Thermo Scientific Chemicals Supplier No. B2290206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 291-64-5 | C7H14 | 98.19 g/mol

Cycloheptane is used as a nonpolar solvent for the chemical industry and as an intermediate in the manufacture of chemicals and pharmaceutical drugs. It can be used to get fluorocycloheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptane is used as a nonpolar solvent for the chemical industry and as an intermediate in the manufacture of chemicals and pharmaceutical drugs. It can be used to get fluorocycloheptane.

Solubility
Soluble in water 30 mg @ 20°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 291-64-5
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00004147
InChI Key DMEGYFMYUHOHGS-UHFFFAOYSA-N
Synonym heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
PubChem CID 9265
IUPAC Name cycloheptane
SMILES C1CCCCCC1

Specifications

Density 0.811
Boiling Point 118°C to 119°C
Flash Point 6°C (43°F)
Refractive Index 1.445
Quantity 5 g
UN Number UN1206
Solubility Information Soluble in water 30 mg @ 20°C.
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material Cycloheptane
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber 206-030-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only