missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecene (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. c046425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C046425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C046425G Supplier TCI America Supplier No. C046425G
Only null left

Chemical Identifiers

CAS 1501-82-2
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003721
InChI Key HYPABJGVBDSCIT-UPHRSURJSA-N
Synonym e-cyclododecene, cyclododecene, e, trans-cyclododecene, 1e-cyclododecene, t-cyclododecene, cyclododecene, trans, 11e-cyclododecene
PubChem CID 5365627
IUPAC Name (Z)-cyclododecene
SMILES C1CCCCC\C=C/CCCC1

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 g
Formula Weight 166.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclododecene (cis- and trans- mixture)
EINECSNumber (3)-2241
TSCA No