missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclododecane 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 63°C |
| Color | White |
| Boiling Point | 111°C |
| Quantity | 500 g |
| Formula Weight | 168.32 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cyclododecane |
Safety and Handling
| EINECSNumber | (3)-2240 |
| RTECSNumber | GV2295000 |
| TSCA | Yes |