missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecane 99.0+%, TCI America™

Catalog No. C055425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C055425G 25 g
C0554100G 100 g
C0554500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C055425G Supplier TCI America Supplier No. C055425G
Only null left

Chemical Identifiers

CAS 294-62-2
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00014258
InChI Key DDTBPAQBQHZRDW-UHFFFAOYSA-N
PubChem CID 9268
IUPAC Name cyclododecane
SMILES C1CCCCCCCCCCC1

Specifications

Melting Point 63°C
Color White
Boiling Point 111°C
Quantity 25 g
Formula Weight 168.32
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclododecane
EINECSNumber (3)-2240
RTECSNumber GV2295000
TSCA Yes