missing translation for 'onlineSavingsMsg'
Learn More

gamma-Cyclodextrin hydrate, 99%

CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol

Supplier:  Thermo Scientific Chemicals 229900050

Quantity 5 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC229900050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Chemical Identifiers

91464-90-3
1297.13
GDSRMADSINPKSL-UHFFFAOYNA-N
71311509
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
C48H80O40
MFCD00149574
gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade
5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol

Specifications

γ-Cyclodextrin hydrate
267.0°C
Authentic
Glass bottle
5 g
14, 2718
+ 177.00
Solubility in water: 22.3 g/100 mL (25°C).
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
1297.13
1297.15
Powder
91464-90-3
White
98.5% min. (ELSD) (HPLC)
C48H80O40
MFCD00149574
+ 177.00 (20.00°C c=1,H2O)
gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade
GDSRMADSINPKSL-UHFFFAOYNA-N
5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
71311509
99%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions