missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Cyclodextrin hydrate, 99%
CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol
Supplier: Thermo Scientific Chemicals 229900010
| Quantity | 1 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
| 91464-90-3 | |
| 1297.13 | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| 71311509 | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C48H80O40 | |
| MFCD00149574 | |
| gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol |
Specifications
| γ-Cyclodextrin hydrate | |
| 267.0°C | |
| Authentic | |
| Glass bottle | |
| 1 g | |
| 14, 2718 | |
| gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol | |
| 71311509 | |
| 99% |
| 91464-90-3 | |
| White | |
| 98.5% min. (ELSD) (HPLC) | |
| C48H80O40 | |
| MFCD00149574 | |
| + 177.00 | |
| Solubility in water: 22.3 g/100 mL (25°C). | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1297.13 | |
| 1297.15 | |
| Powder |