missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Cyclodextrin hydrate, 99%
CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol
Supplier: Thermo Scientific Chemicals 227280250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
| 68168-23-0 | |
| 1134.99 | |
| WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
| 53486154 | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C42H70O35 | |
| MFCD00150811 | |
| beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
| 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol |
Specifications
| β-Cyclodextrin hydrate | |
| 68168-23-0 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| 25 g | |
| 06,151; 08,133; 09,129; 12,151; 15,107 | |
| + 161 | |
| (1% in water) Clear colorless | |
| 5ppm toluene max. | |
| 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol | |
| 53486154 | |
| 99% | |
| Crystalline Powder |
| 99% | |
| 98.5 | |
| 298°C to 300°C | |
| 5.0 to 8.0 (1% aq. soln.) | |
| 98.5% min. (ELSD) (HPLC) | |
| C42H70O35 | |
| MFCD00150811 | |
| 14, 2718 | |
| beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc | |
| WHGYBXFWUBPSRW-UHFFFAOYSA-N | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1134.99 | |
| 1135.01 | |
| 1% max. Alpha-and gamma-cyclodextrin (HPLC) |
Safety and Handling
TSCA : TSCA