missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin, 98%

Catalog No. AC406000250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC406000250 25 g
AC406001000 100 g
AC406005000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC406000250 Supplier Thermo Scientific Chemicals Supplier No. 406000250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 310.0°C
Loss on Drying 14% max. (105°C, 8 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Quantity 25 g
Fieser 06,151; 08,133; 09,129; 12,151; 15,107
Merck Index 15,271
Specific Rotation Condition + 150.00 (25.00°C c=1.5, in water)
Specific Rotation + 150.00
Solubility Information Solubility in water: soluble.
Formula Weight 1134.98
Percent Purity 98%
Chemical Name or Material β-Cyclodextrin
Show More Show Less
EINECSNumber 231-493-2

RUO – Research Use Only