missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine, 98%

CAS: 2516-34-9 | C4H9N | 71.123 g/mol

Supplier:  Thermo Scientific Chemicals A1342314

 View more versions of this product

Catalog No. AAA1342314


Only null left
Explore available promotions

Description

Description

Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
Specifications

Specifications

Cyclobutylamine
0.836
−4°C (24°F)
1.4376
25 g
2069297
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
KZZKOVLJUKWSKX-UHFFFAOYSA-N
cyclobutanamine
75645
98%
2516-34-9
82°C to 83°C
C4H9N
MFCD00001328
UN2733
Air sensitive; Hygroscopic
Slightly soluble in water.
C1CC(C1)N
71.123
71.12

Chemical Identifiers

2516-34-9
71.123
KZZKOVLJUKWSKX-UHFFFAOYSA-N
75645
C1CC(C1)N
C4H9N
MFCD00001328
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
cyclobutanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 219-736-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only