missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine, 98%

Catalog No. AAA1342303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1342303 1 g
AAA1342306 5 g
AAA1342314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1342303 Supplier Thermo Scientific Chemicals Supplier No. A1342303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2516-34-9 | C4H9N | 71.123 g/mol

Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 2516-34-9
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00001328
InChI Key KZZKOVLJUKWSKX-UHFFFAOYSA-N
Synonym cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
PubChem CID 75645
IUPAC Name cyclobutanamine
SMILES C1CC(C1)N

Specifications

Density 0.836
Boiling Point 82°C to 83°C
Flash Point −4°C (24°F)
Refractive Index 1.4376
Quantity 1 g
UN Number UN2733
Beilstein 2069297
Sensitivity Air sensitive; Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 71.12
Percent Purity 98%
Chemical Name or Material Cyclobutylamine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 219-736-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only