missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanone (stabilized with Na2CO3) 97.0+%, TCI America™

Catalog No. C19131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19131G 1 g
C19135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19131G Supplier TCI America Supplier No. C19131G

Chemical Identifiers

CAS 1191-95-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00001332
InChI Key SHQSVMDWKBRBGB-UHFFFAOYSA-N
Synonym unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
PubChem CID 14496
IUPAC Name cyclobutanone
SMILES O=C1CCC1

Specifications

Melting Point -53°C
Color Yellow
Boiling Point 99°C
Quantity 1 g
UN Number 1224
Formula Weight 70.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclobutanone (stabilized with Na2CO3)
TSCA Yes
Recommended Storage Freezer