missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanone, 98%, stab. with ca 0.1% BHT

Catalog No. AAA1306803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1306803 1 g
AAA1306806 5 g
AAA1306814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1306803 Supplier Thermo Scientific Chemicals Supplier No. A1306803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1191-95-3 | C4H6O | 70.09 g/mol

Cyclobutanone is used in preparation of cyclobutane derivatives. It is used as a starting material for aminocyclobutanecarboxylic acid through sulfonyloxiranes. It is also used to prepare dihydro-furan-2-one in presence of polystyrene-bound phenylselenic acid as catalyst. Further, it is involved in the photochemical synthesis of nucleoside analogues such as bicyclic nucleosides and isonucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutanone is used in preparation of cyclobutane derivatives. It is used as a starting material for aminocyclobutanecarboxylic acid through sulfonyloxiranes. It is also used to prepare dihydro-furan-2-one in presence of polystyrene-bound phenylselenic acid as catalyst. Further, it is involved in the photochemical synthesis of nucleoside analogues such as bicyclic nucleosides and isonucleosides.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

Chemical Identifiers

CAS 1191-95-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00001332
InChI Key SHQSVMDWKBRBGB-UHFFFAOYSA-N
Synonym unii-6pf2sh405u, cyclobutyloxy, cyclobutanone, 98+%, cyclobutanon, cylcobutanone, cylobutanone, 3-cyclobutanone, 1-oxocyclobutane, cyclobutanone, pubchem9021
PubChem CID 14496
IUPAC Name cyclobutanone
SMILES O=C1CCC1

Specifications

Melting Point -51°C
Density 0.935
Boiling Point 97°C to 99°C
Flash Point 10°C (50°F)
Odor Pleasant
Refractive Index 1.421
Quantity 1 g
UN Number UN1224
Beilstein 1560289
Sensitivity May form a precipitate
Solubility Information Immiscible with water.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclobutanone, stabilized with ≈0.01% BHT
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 214-745-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only