missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanol 98.0+%, TCI America™

Catalog No. p-7124598
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C07701G 1 g
C07705G 5 g
2 options

Catalog No. C07701G

Supplier: TCI America C07701G

Only null left

Chemical Identifiers

CAS 2919-23-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001318
InChI Key KTHXBEHDVMTNOH-UHFFFAOYSA-N
Synonym cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
PubChem CID 76218
IUPAC Name cyclobutanol
SMILES OC1CCC1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
UN Number 1987
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclobutanol