missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanol, 99+%

Catalog No. AC168750010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168750010 1 g Glass bottle
AC168750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168750010 Supplier Thermo Scientific Chemicals Supplier No. 168750010

CAS: 2919-23-5 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2919-23-5
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001318
InChI Key KTHXBEHDVMTNOH-UHFFFAOYSA-N
Synonym cyclobutyl alcohol, cyclobutyl hydroxide, hydroxycyclobutane, acmc-1cgim, cyclobutanol, cyclobutanol 1g, ksc206s6t, kthxbehdvmtnoh-uhfffaoysa
PubChem CID 76218
IUPAC Name cyclobutanol
SMILES OC1CCC1

Specifications

Color Colorless
Density 0.9200g/mL
Boiling Point 122.0°C to 123.0°C (733.0 mmHg)
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370
Linear Formula C4H7OH
Quantity 1 g
Beilstein 06, 4
Specific Gravity 0.92
Formula Weight 72.11
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Cyclobutanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-858-1

RUO – Research Use Only