missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic Acid 97.0+%, TCI America™

Catalog No. C088825G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088825G 25 g
C088810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C088825G Supplier TCI America Supplier No. C088825G

Chemical Identifiers

CAS 3721-95-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001323
InChI Key TXWOGHSRPAYOML-UHFFFAOYSA-N
Synonym cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acid
PubChem CID 19494
IUPAC Name cyclobutanecarboxylic acid
SMILES OC(=O)C1CCC1

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 g
UN Number 3265
Formula Weight 100.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic Acid
RTECSNumber GU1335000
TSCA Yes