missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid chloride, 99%

Catalog No. AC110910010
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110910010 1 g Glass bottle
AC110910100 10 g Glass bottle
AC110910500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110910010 Supplier Thermo Scientific Chemicals Supplier No. 110910010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl

Specifications

Color Colorless
Density 37g/mL
Boiling Point 145.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4545 to 1.4565
Linear Formula C4H7COCl
Quantity 1 g
Beilstein 09, 5
Formula Weight 118.56
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-680-8

RUO – Research Use Only