missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid, 98%

Catalog No. AC110901000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110901000 100 g Glass bottle
AC110900050 5 g Glass bottle
AC110900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110901000 Supplier Thermo Scientific Chemicals Supplier No. 110901000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3721-95-7 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3721-95-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001323
InChI Key TXWOGHSRPAYOML-UHFFFAOYSA-N
Synonym cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acid
PubChem CID 19494
IUPAC Name cyclobutanecarboxylic acid
SMILES OC(=O)C1CCC1

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 195.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4410 to 1.4450
Linear Formula C4H7CO2H
Quantity 100 g
Beilstein 09, 5
Specific Gravity 1.04
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-072-7

RUO – Research Use Only