missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid, 97%

Catalog No. p-7021939
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1448014 25 g
AAA1448022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1448014 Supplier Thermo Scientific Chemicals Supplier No. A1448014
Only null left

CAS: 3721-95-7 | C5H8O2 | 100.12 g/mol

Cyclobutanecarboxylic acid is used in water treatment, row materials for sodium fluosilicate potassium fluosilicateammonium fluosilicatecopper fluosilicatebarium fluosilicate and other fluosilicates and silicic tetrafluoride. It is also used in refining lead of electrolysis, as mordant and in the treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutanecarboxylic acid is used in water treatment ,row materials for sodium fluosilicate potassium fluosilicateammonium fluosilicatecopper fluosilicatebarium fluosilicate and other fluosilicates and silicic tetrafluoride. It is also used in metal platingtimber antisepsisdisinfectant(1-2% of solution) of beer and brewing industry, refining lead of electrolysis, as mordant and in the treatment of metal surface.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS 3721-95-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001323
InChI Key TXWOGHSRPAYOML-UHFFFAOYSA-N
Synonym cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acid
PubChem CID 19494
IUPAC Name cyclobutanecarboxylic acid
SMILES OC(=O)C1CCC1

Specifications

Melting Point -20°C
Density 1.055
Boiling Point 194°C to 195°C
Flash Point 83°C (181°F)
Odor Stench
Refractive Index 1.4433
Quantity 25 g
UN Number UN3265
Beilstein 1816777
Solubility Information Slightly soluble in water.
Formula Weight 100.12
Percent Purity ≥98%
Chemical Name or Material Cyclobutanecarboxylic acid
Show More Show Less
Hazard Category H227-H302-H312-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-072-7
RTECSNumber GU1335000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only