missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarbonyl chloride, 98%

Catalog No. AAB2031709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2031709 10 g
AAB2031718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2031709 Supplier Thermo Scientific Chemicals Supplier No. B2031709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl

Specifications

Density 1.039
Boiling Point 145°C
Flash Point 37°C (99°F)
Odor Stench
Refractive Index 1.455
Quantity 10 g
UN Number UN2920
Beilstein 956624
Sensitivity Moisture Sensitive
Formula Weight 118.56
Percent Purity 98%
Chemical Name or Material Cyclobutanecarbonyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 225-680-8
TSCA No

RUO – Research Use Only