missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclobis(paraquat-1,4-phenylene) Tetrakis(hexafluorophosphate) 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 275°C |
| Quantity | 100 mg |
| Formula Weight | 1100.54 |
| Percent Purity | ≥94.0% (HPLC) |
| Grade | Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cyclobis(paraquat-1,4-phenylene) Tetrakis(hexafluorophosphate) |
Safety and Handling
| TSCA | No |