missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric Acid 98.0+%, TCI America™

Catalog No. C0459500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0459500G 500 g
C045925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0459500G Supplier TCI America Supplier No. C0459500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1

Specifications

Melting Point 360°C
Color White
Quantity 500 g
Physical Form Crystalline Powder
Formula Weight 129.08
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Cyanuric Acid
EINECSNumber (5)-1037&(5)-1038
RTECSNumber XZ1800000
TSCA Yes