missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanopyrazine 99.0+%, TCI America™
$249.78 - $249.78
Chemical Identifiers
| CAS | 19847-12-2 |
|---|---|
| Molecular Formula | C5H3N3 |
| Molecular Weight (g/mol) | 105.10 |
| MDL Number | MFCD00049361 |
| InChI Key | PMSVVUSIPKHUMT-UHFFFAOYSA-N |
| Synonym | pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine |
| PubChem CID | 73172 |
| ChEBI | CHEBI:3982 |
| IUPAC Name | pyrazine-2-carbonitrile |
| SMILES | N#CC1=CN=CC=N1 |
Chemical Identifiers
| 19847-12-2 | |
| 105.10 | |
| PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
| 73172 | |
| pyrazine-2-carbonitrile |
| C5H3N3 | |
| MFCD00049361 | |
| pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
| CHEBI:3982 | |
| N#CC1=CN=CC=N1 |
Specifications
| 19847-12-2 | |
| 87°C | |
| MFCD00049361 | |
| pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine | |
| N#CC1=CN=CC=N1 | |
| 105.10 | |
| CHEBI:3982 | |
| ≥99.0% (GC) | |
| Cyanopyrazine |
| White | |
| C5H3N3 | |
| 500 g | |
| PMSVVUSIPKHUMT-UHFFFAOYSA-N | |
| pyrazine-2-carbonitrile | |
| 73172 | |
| 105.10 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (5)-5849
TSCA : Yes