missing translation for 'onlineSavingsMsg'
Learn More

Cyanopyrazine 99.0+%, TCI America™

Catalog No. C1123500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1123500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1123500G Supplier TCI America Supplier No. C1123500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19847-12-2
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00049361
InChI Key PMSVVUSIPKHUMT-UHFFFAOYSA-N
Synonym pyrazinecarbonitrile, 2-cyanopyrazine, cyanopyrazine, 2-pyrazinecarbonitrile, pyrazinenitrile, pyrazinonitrile, 2-cyanopyarine, 2-cyano pyrazine
PubChem CID 73172
ChEBI CHEBI:3982
IUPAC Name pyrazine-2-carbonitrile
SMILES N#CC1=CN=CC=N1

Specifications

Color White
Boiling Point 87°C
Quantity 500 g
Formula Weight 105.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Cyanopyrazine
EINECSNumber (5)-5849
TSCA Yes