missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen bromide

Catalog No. AA3273106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3273106 5 g
AA3273114 25 g
AA3273122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3273106 Supplier Thermo Scientific Chemicals Supplier No. 03273106

CAS: 506-68-3 | CBrN | 105.922 g/mol

Cyanogen bromide is used as a reagent in molecular biology, peptide mapping, gold extraction and amino acid sequence analysis. It acts as a coupling reagent for phosphoramidate or pyrophosphate inter-nucleotide bonds in DNA duplexes. It is also used in organic synthesis to prepare cyanamides, dicyanamides, cyanuric bromide, guanidines and hydroxylguanidines. It is involved in the von Braun reaction, it gives disubstituted cyanamides by reacting with tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 506-68-3
Molecular Formula CBrN
Molecular Weight (g/mol) 105.922
MDL Number MFCD00011597
InChI Key ATDGTVJJHBUTRL-UHFFFAOYSA-N
Synonym cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
PubChem CID 10476
IUPAC Name carbononitridic bromide
SMILES C(#N)Br

Specifications

Melting Point 50°C to 52°C
Density 2.015
Boiling Point 61.4°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethanol,diethyl ether,benzene and acetonitrile.
Formula Weight 105.93
Physical Form Crystals
Chemical Name or Material Cyanogen bromide
Hazard Category H300+H310+H330-H314-H335-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 208-051-2
RTECSNumber GT2100000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only