Learn More
Cyanogen bromide, 97%
CAS: 506-68-3 | CBrN | 105.922 g/mol
$118.00 - $118.00
Chemical Identifiers
| CAS | 506-68-3 |
|---|---|
| Molecular Formula | CBrN |
| MDL Number | MFCD00011597 |
| InChI Key | ATDGTVJJHBUTRL-UHFFFAOYSA-N |
| Synonym | cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn |
| PubChem CID | 10476 |
| IUPAC Name | carbononitridic bromide |
| SMILES | C(#N)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC110780250
|
Thermo Scientific Chemicals
110780250 |
25 g | Glass Bottle in Tin Can |
Each for $118.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
| 506-68-3 | |
| MFCD00011597 | |
| cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn | |
| carbononitridic bromide |
| CBrN | |
| ATDGTVJJHBUTRL-UHFFFAOYSA-N | |
| 10476 | |
| C(#N)Br |
Specifications
| 506-68-3 | |
| 1.44 | |
| Glass Bottle in Tin Can | |
| BrCN | |
| 25 g | |
| 15, 2683 | |
| ATDGTVJJHBUTRL-UHFFFAOYSA-N | |
| carbononitridic bromide | |
| 10476 | |
| ≥96.0% | |
| Cyanogen bromide, 97% |
| White | |
| 97% | |
| CBrN | |
| MFCD00011597 | |
| 01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127 | |
| cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn | |
| C(#N)Br | |
| 105.922 | |
| 105.93 | |
| Crystals |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only