missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Cyanogen bromide

Catalog No. C91492100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C91492100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C91492100G Supplier Sigma Aldrich Supplier No. C91492100G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyanogen bromide

Specifications

Boiling Point 61°C to 62°C (lit.)
Melting Point 50°C to 53°C (lit.)
CAS 506-68-3
Quantity 100 g
Molecular Formula CBrN
Linear Formula BrCN
MDL Number MFCD00011597
Synonym Bromine cyanide
Molecular Weight (g/mol) 105.92
Percent Purity 97%