missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetic acid, 97%

Catalog No. AC110670250
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110670250 25 g Glass bottle
AC110672500 250 g Glass bottle
AC110670010 1 kg Glass bottle
AC110670050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC110670250 Supplier Thermo Scientific Chemicals Supplier No. 110670250

CAS: 372-09-8 | C3H3NO2 | 85.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-09-8
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.06
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
Synonym cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
PubChem CID 9740
ChEBI CHEBI:51889
IUPAC Name 2-cyanoacetic acid
SMILES C(C#N)C(=O)O

Specifications

Melting Point 65.0°C to 67.0°C
Color White to Beige
Boiling Point 108.0°C (15.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCCH2CO2H
Quantity 25 g
Beilstein 02,583
Fieser 11,145; 13,171
Merck Index 15,2679
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene and chloroform
Formula Weight 85.06
Percent Purity 97%
Physical Form Adhering Crystalline Solid
Chemical Name or Material Cyanoacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 206-743-9

RUO – Research Use Only