missing translation for 'onlineSavingsMsg'
Learn More

Cyanine 98.0+%, TCI America™

Catalog No. C04361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04361G 1 g
C04365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04361G Supplier TCI America Supplier No. C04361G
Only null left

Chemical Identifiers

CAS 523-42-2
Molecular Formula C29H35IN2
Molecular Weight (g/mol) 538.517
MDL Number MFCD00050330
InChI Key QGKMIGUHVLGJBR-UHFFFAOYSA-M
Synonym Quinoline Blue
PubChem CID 91659094
IUPAC Name (4Z)-1-(3-methylbutyl)-4-[[1-(3-methylbutyl)quinolin-1-ium-4-yl]methylidene]quinoline;iodide
SMILES CC(C)CCN1C=CC(=CC2=CC=[N+](C3=CC=CC=C23)CCC(C)C)C4=CC=CC=C41.[I-]

Specifications

Quantity 1 g
Formula Weight 538.52
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyanine