missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide, 95%, stabilized

Catalog No. p-2691249
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181950250 Supplier Thermo Scientific Chemicals Supplier No. 181950250
Only null left

CAS: 420-04-2 | CH2N2 | 42.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 420-04-2
Molecular Formula CH2N2
Molecular Weight (g/mol) 42.04
MDL Number MFCD00007572
InChI Key XZMCDFZZKTWFGF-UHFFFAOYSA-N
Synonym hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
PubChem CID 9864
ChEBI CHEBI:16698
IUPAC Name cyanamide
SMILES C(#N)N

Specifications

Melting Point 40.0°C to 47.0°C
Color White
Boiling Point 260.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NCN
Quantity 25 g
Beilstein 03, IV, 145
Fieser 01,170; 03,71; 05,168; 12,147
Merck Index 15, 2674
Solubility Information Solubility in water: 775g/L. Other solubilities: soluble in alchol,phenols,amines,ethers,ketones,slightly soluble in benzene and halogenated,hydrocarbons
Stabilizer 0.1% (typically) formic acid
Formula Weight 42.04
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material Cyanamide, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Suspecte
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 206-992-3
Recommended Storage Refrigerator +4°C

RUO – Research Use Only