missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CXCR4 Antibody Blocking Peptide

Catalog No. NB398710
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398710 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398710 Supplier R&D Systems™ Supplier No. NB10056437PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Gene ID (Entrez) 7852
Product Line Novus
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-56437
Gene Alias CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor
Gene Symbol CXCR4
Quantity 0.1 mg
Research Category Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells
Storage Buffer Peptide dissolved in dH20. Contains no BSA.