Learn More
Mettler Toledo™ Cuvette Holder
Catalog No. 01911525
Change view
Click to view available options
Pathlength:
10/16 mm
20/30 mm
40/50 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pathlength
01911525 40/50 mm
01911523 10/16 mm
01911524 20/30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01911525 Supplier Mettler Toledo™ Supplier No. 30705124
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mettler Toledo cuvette holders provide a secure base for your sample, ensuring highly accurate measurement results.
  • Compatible with Easy UV and Easy Vis Spectrophotometers
Specifications
Type Cell Holder System
Pathlength 40/50 mm