missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Cuvette for Cobas Mira, case of 300

Catalog No. 361016817
Change view
Click to view available options
Quantity:
300/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361016817 300/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361016817

Designed for use on the Cobas Mira, Mira S and Mira Plus analyzers. Globe Scientific Cuvettes for Cobas Mira come individually wrapped and are constructed of polymethylmethacrylate. These 12-place Cuvette segments are designed for compatibility with these products. Case of 300.

  • For use on the Cobas Mira, Mira S and Mira Plus analyzers
  • 12-place cuvette segments
  • Individually wrapped
  • Polymethylmethacrylate (PMMA)
  • Natural color
  • Case of 300

Specifications

For Use With (Equipment) Cobas Mira, Mira S, and Mira Plus Analyzers
Color Natural
Quantity 300/Cs.
Material PMMA