Learn More
Globe Scientific Cuvette for Cobas Mira, case of 500
Designed for use on the Cobas Mira, Mira S and Mira Plus analyzers. Globe Scientific Cuvettes for Cobas Mira come individually wrapped and are constructed of polymethylmethacrylate. These 12-place Cuvette segments are designed for compatibility with these products. Case of 500.
Supplier: Globe Scientific 5120
Description
- For use on the Cobas Mira, Mira S and Mira Plus analyzers
- 12-place cuvette segments
- Individually wrapped
- Polymethylmethacrylate (PMMA)
- Natural color
- Case of 500
Specifications
| Cuvette | |
| PMMA | |
| Cobas Mira, Mira S, and Mira Plus analyzers |
| Natural color | |
| Case of 500, individually wrapped |