Learn More
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath 10mm)
Catalog No. 501059831
Encompass
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence work Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL
Catalog No. 501059831
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 9FL Semi-Micro Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence work Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL