Learn More
Type 9B Semi-Micro Raised-Bottom Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10x1mm)
Catalog No. 501059818
Encompass
Type 9B Semi-Micro Raised-Bottom Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10x1mm) Cuvette has an open top with a PTFE lid which rests on the top Cuvette has a 9mm raised bottom Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.4mL
Catalog No. 501059818
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 9B Semi-Micro Raised-Bottom Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10x1mm) Cuvette has an open top with a PTFE lid which rests on the top Cuvette has a 9mm raised bottom Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 10x1mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x1x45mm Capacity- 0.4mL