Learn More
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm)
Catalog No. 501059802
Encompass
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL
Catalog No. 501059802
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 9 Semi-Micro Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL