Learn More
Type 82 Triangular Fluorescence Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm)
Catalog No. 501060311
Encompass
Type 82 Triangular Fluorescence Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm) Cuvette has an airtight PTFE stopper which plugs into the top Cuvette has a triangular shape Material is UV Quartz (transmits over 80% between 190-2,500nm) Three windows and the base optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x49mm Inside Dimensions- 10x10x45mm Capacity- 1.75mL
Catalog No. 501060311
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 82 Triangular Fluorescence Cell with PTFE Stopper (Material: UV Quartz) (Lightpath: 10mm) Cuvette has an airtight PTFE stopper which plugs into the top Cuvette has a triangular shape Material is UV Quartz (transmits over 80% between 190-2,500nm) Three windows and the base optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x49mm Inside Dimensions- 10x10x45mm Capacity- 1.75mL