Learn More
Type 700 Disposable Sub-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic) (Z-Dimension: 8.5mm)
Catalog No. 501060451
Encompass
Type 700 Disposable Sub-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic) (Z-Dimension: 8.5mm) Cuvette has an open top with optional push caps that fit into the top (part # for the caps is D020) Comes in a box of 100 Material is UV Plastic (transmits over 80% between 220-900nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x2x45mm z-dimension- 8.5mm Chamber height- 3.5mm Capacity- 70L
Catalog No. 501060451
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 700 Disposable Sub-Micro Cuvettes (Lightpath: 10mm) (Material: UV Plastic) (Z-Dimension: 8.5mm) Cuvette has an open top with optional push caps that fit into the top (part # for the caps is D020) Comes in a box of 100 Material is UV Plastic (transmits over 80% between 220-900nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x2x45mm z-dimension- 8.5mm Chamber height- 3.5mm Capacity- 70L