Learn More
Type 61FL Macro Graded Seal Fluorescence Tube Cuvette (Material: UV Quartz) (Lightpath: 10mm)
Catalog No. 501060290
Encompass
Type 61FL Macro Graded Seal Fluorescence Tube Cuvette (Material: UV Quartz) (Lightpath: 10mm) Cuvette has a graded seal tube on top with a quartz-to-pyrex bond. Additional Pyrex Glass structures can be fused to the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x125mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL
Catalog No. 501060290
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 61FL Macro Graded Seal Fluorescence Tube Cuvette (Material: UV Quartz) (Lightpath: 10mm) Cuvette has a graded seal tube on top with a quartz-to-pyrex bond. Additional Pyrex Glass structures can be fused to the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x125mm Inside Dimensions- 10x10x45mm Capacity- 3.5mL