Learn More
Type 507 Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 3x3x40mm)
Catalog No. 501060346
Encompass
Type 507 Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 3x3x40mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 3x3x40mm Outside dimensions 5.5x5.5x40mm Inside Dimensions- 3x3x40mm Capacity- 0.28mL
Catalog No. 501060346
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 507 Micro Fluorescence Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 3x3x40mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 3x3x40mm Outside dimensions 5.5x5.5x40mm Inside Dimensions- 3x3x40mm Capacity- 0.28mL