Type 46 Semi-Micro Cuvette with Screw Cap (Material: UV Quartz) (Lightpath: 10x4mm) Cuvette has an airtight cap, which screws onto the top of the cuvette Each cell comes standard with one closed screw cap and one open screw cap with a self-healing injection disc (SEPTA) Material is UV Quartz (transmits over 80% between 190-2,500nm) Two windows optically polished Lightpath is 10x4mm Outside dimensions 12.5x12.5x55mm Inside Dimensions- 10x4x45mm Capacity- 1.4mL