Type 4 Micro Fluorescence Cuvette with PTFE Cover (Material: UV Quartz) (Lightpath 5X5X33mm) Cuvette has an open top with a PTFE lid which rests on the top Material is UV Quartz (transmits over 80% between 190-2,500nm) Four windows and the base optically polished for fluorescence measurements Lightpath is 5x5x33mm Outside dimensions 7.5x7.5x33mm Inside Dimensions- 3x3x40mm Capacity- 0.64mL. *Does not come with the black holder/adapter.*