Learn More
Type 18 Standard Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm)
Catalog No. 501059923
Encompass
Type 18 Standard Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x2x45mm Capacity- 0.7mL
Catalog No. 501059923
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 18 Standard Micro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 10mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Two windows optically polished Lightpath is 10mm Outside dimensions 12.5x12.5x45mm Inside Dimensions- 10x2x45mm Capacity- 0.7mL