Learn More
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 1mm)
Catalog No. 501059733
Encompass
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 1mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Two windows optically polished Lightpath is 1mm Outside dimensions 3.5x12.5x45mm Inside Dimensions- 1x10x45mm Capacity- 0.4mL
Catalog No. 501059733
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Type 1 Macro Cuvette with PTFE Cover (Material: Optical Glass) (Lightpath: 1mm) Cuvette has an open top with a PTFE lid which rests on the top Material is Optical Glass (transmits over 80% between 340-2,500nm) Two windows optically polished Lightpath is 1mm Outside dimensions 3.5x12.5x45mm Inside Dimensions- 1x10x45mm Capacity- 0.4mL