missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
$1,218.28
Specifications
| Sample Type | Undiluted Serum |
|---|---|
| For Use With (Equipment) | Reichert Unistat™ Bilirubinometer |
| Capacity (Metric) | 20 μL |
Description
- For use with Unistat Bilirubinometer
- Capacity: 20μL
Specifications
| Undiluted Serum | |
| Reichert Unistat™ Bilirubinometer | |
| 20 μL |