missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves

Catalog No. 17200350
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17200350 Small
17200351 Medium
17200352 Large
17200353 X-Large
17200354 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17200350 Supplier Wells Lamont™ Supplier No. I2469S

FlexTech™ Cool Series Nitrile-Coated Anti-Impact Gloves feature soft and flexible TPR pads that provide an additional layer of protection against impacts to the back of the hand and fingers. Nitrile coating on the palm improves grip capabilities when handling wet and oily parts.

  • A blend of high performance and cooling fibers extends comfort in warmer work environments
  • Soft and flexible TPR pads provide an additional layer of protection against blows to the back of the hand and fingers
  • Reinforced thumb crotch for extra protection between the thumb and forefinger
  • An ANSI Cut Level A4 ensures a high level of cut protection against sharp objects
  • Padded, sandy nitrile coated palm increases protection and improves grip
  • Ideal for handling parts in wet and oily environments

Specifications

ANSI Abrasion Rating ANSI 4
Applications Industrial
Coating Material Nitrile
Color Hi-vis Orange
Cut Resistance Rating ANSI A4
Exterior Finish Fully Textured
Hand Specific
Material HPPE
Product Type Anti-impact Gloves
Size Small
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 4
Resistant To Cuts, Abrasions, Impacts, Punctures
Series I2469
Tear Rating 4
Yarn Gauge 15
Show More Show Less