missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Custom Black Phenolic Work Surface

Catalog No. 06000387
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
06000387 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06000387

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Custom Phenolic Work Surface is based on units of work surface designed and fabricated to suit your needs.

  • Contact your Fisher representative for details
  • 1 in. thick black phenolic

Specifications

Product Type Work Surface
Height (English) Phenolic Work Surface
For Use With (Application) Work Surface
Includes Work Surface
No. of Doors None
No. of Drawers None
Warranty 2 Years
Color Black
Height (Metric) 2.5 cm
Material Phenolic