Learn More
NoTrax™ Cushion Trax™ Anti-Fatigue Mat
GREENER_CHOICE
Cushion Trax™ is an anti-fatigue floor mat with a heavy-duty vinyl diamond-plate design and a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.
Supplier:  NoTrax™ 479R4875YB
 View more versions of this product
Catalog No. 19015798
Only null left
Description
Description
  • Cushion Trax™ is engineered to meet the toughest requirements of today's industrial applications
  • It is particularly suited for multi-shift operations in dry work areas where the worker is standing for prolonged periods of time such as workstations and assembly lines
  • The heavy-duty vinyl diamond-plate design provides non-directional traction, and is easy to clean
  • The top surface is combined with a dense closed cell foam base utilizing NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
  • NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate the slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
  • The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat
Specifications
Chemical Identifiers
Certified by the NFSI (National Floor Safety Institute)
Anti-fatigue Floor Mat
9/16 in.
Nitricell, PVC
Manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual workstations. Suitable for commercial environments including hotels, reception desks, shop counters, department stores, banks, restaurants, etc.
14.2875 mm
Specifications
Manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual workstations. Suitable for commercial environments including hotels, reception desks, shop counters, department stores, banks, restaurants, etc.
Certified by the NFSI (National Floor Safety Institute)
Black/Yellow Striped
75 ft.
4 ft.
1.2192 m
Nitricell, PVC
9/16 in.
Anti-fatigue Floor Mat
75 x 4 ft.
22.86 m
14.2875 mm
Product Suggestions
Product Suggestions